Order Entry
Belgium
ContactUsLinkComponent
Sodium carbonate, anhydrous 99.95-100.05%, granular, AR® ACS primary standard, Macron Fine Chemicals™
Irritant
Sodium carbonate, anhydrous 99.95-100.05%, granular, AR® ACS primary standard, Macron Fine Chemicals™
Certificates
undefined
Irritant
Sodium carbonate, anhydrous 99.95-100.05%, granular, AR® ACS primary standard, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (Na₂CO₃ (after 2 hr @ 285°C)):
    99.95 - 100.05 %
  • ACS - Calcium (Ca):
    ≤0.02 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.0005 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.0005 %
  • ACS - Loss on Heating at 285°C:
    ≤1.0 %
  • ACS - Magnesium (Mg):
    ≤0.004 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Silica (SiO₂):
    ≤0.005 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.003 %
  • ACS - Solubility (8 in 50):
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together