Order Entry
Belgium
ContactUsLinkComponent
Sodium acetate trihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium acetate trihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium acetate trihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 101.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (CH₃COONa·3H₂O):
    99.0 - 101.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    7.5 - 9.2
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Substances Reducing Permanganate:
    Passes Test
  • Potassium (K):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together