Order Entry
Belgium
ContactUsLinkComponent
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, Multi-Compendial endotoxin-free, J.T.Baker®
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, Multi-Compendial endotoxin-free, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
di-Potassium hydrogen phosphate 98.0-100.5% (dried basis) USP, Multi-Compendial endotoxin-free, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤3 ppm
  • Meets B.P. Chemical Specifications:
    Passes Test
  • Meets E.P. Chemical Specifications:
    ≤0.03%
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.001%
  • USP - Assay (K₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤ 3 ppm
  • USP - Carbonate (CO₃):
    Passes Test
  • USP - Chloride (Cl):
    ≤ 0.03 %
  • USP - Endotoxin Concentration (2.5 EU/g max.):
    Passes Test
  • USP - Fluoride (F):
    ≤ 0.001 %
  • USP - Identification A (USP):
    Passes Test
  • USP - Identification B (USP):
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.2 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Loss on Drying:
    ≤ 1.0 %
  • USP - Mono- or Tri- Basic Salt:
    Passes Test
  • USP - pH (1 in 20):
    8.5 - 9.6
  • USP - Sodium (Na):
    Passes Test
  • USP - Sulfate (SO₄):
    ≤ 0.1 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Assay (K₂HPO₄) (dried basis):
    98.0 - 101.0 %
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 2.0 %
  • EP/BP - Monopotassium phosphate:
    ≤ 0.025
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Sodium (Na):
    ≤ 0.1 %
  • EP/BP - Sulfate (SO₄):
    ≤ 0.1 %

Specification Test Results

Related Information

Frequently Bought Together