Order Entry
Belgium
ContactUsLinkComponent
Potassium dichromate concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Potassium dichromate concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4671.
Supplier:  Avantor
CAS Number:  
Potassium dichromate concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4671.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    210 mL
  • Seq:
    2545

Specifications

Specification Test Results

  • Titer (N):
    0.0998-0.1002

Specification Test Results

About this item

 :
 :294.18 g/mol
UN:3087
ADR:5.1,II
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367