Order Entry
Belgium
ContactUsLinkComponent
Potassium bromate 0.1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Health
Potassium bromate 0.1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Catalog # 5629.1000
Supplier:  Avantor
CAS Number:  
undefined
Health
Potassium bromate 0.1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Catalog # 5629.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0995 - 0.1005
  • Normality:
    0.0995 - 0.1005

Specification Test Results

About this item

 :
 :167 g/mol
ADR:5.1,II
 :Management of Change (MOC) category = N
EINECS:231-829-8
UN:1484
 :GHS08
Storage Temperature:Ambient
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359