Order Entry
Belgium
ContactUsLinkComponent
Perchloric acid 70%, BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Oxidizing
Corrosive
Irritant
Perchloric acid 70%, BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Corrosive
Irritant
Perchloric acid 70%, BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    65 - 71 %
  • Color (APHA):
    ≤10
  • Assay (HClO₄):
    65 - 71 %
  • Trace Impurities - Aluminum (Al):
    ≤1 ppb
  • Trace Impurities - Antimony (Sb):
    ≤0.5 ppb
  • Trace Impurities - Arsenic (As):
    ≤0.5 ppb
  • Trace Impurities - Barium (Ba):
    ≤1 ppb
  • Trace Impurities - Beryllium (Be):
    ≤0.5 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤0.5 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤1 ppb
  • Trace Impurities - Calcium (Ca):
    ≤1 ppb
  • Trace Impurities - Cerium (Ce):
    ≤0.5 ppb
  • Trace Impurities - Cesium (Cs):
    ≤0.5 ppb
  • Trace Impurities - Cobalt (Co):
    ≤0.5 ppb
  • Trace Impurities - Copper (Cu):
    ≤0.5 ppb
  • Trace Impurities - Dysprosium (Dy):
    ≤0.5 ppb
  • Trace Impurities - Erbium (Er):
    ≤0.5 ppb
  • Trace Impurities - Europium (Eu):
    ≤0.5 ppb
  • Trace Impurities - Gadolinium (Gd):
    ≤0.5 ppb
  • Trace Impurities - Gallium (Ga):
    ≤0.5 ppb
  • Trace Impurities - Gold (Au):
    ≤0.5 ppb
  • Trace Impurities - Hafnium (Hf):
    max. 0.5
  • Trace Impurities - Holmium (Ho):
    ≤0.5 ppb
  • Trace Impurities - Indium (In):
    ≤0.5 ppb
  • Trace Impurities - Iron (Fe):
    ≤1 ppb
  • Trace Impurities - Lanthanum (La):
    ≤0.5 ppb
  • Trace Impurities - Lead (Pb):
    ≤1 ppb
  • Trace Impurities - Lithium (Li):
    ≤0.5 ppb
  • Trace Impurities - Lutetium (Lu):
    ≤0.5 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤1 ppb
  • Trace Impurities - Manganese (Mn):
    ≤1 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤0.5 ppb
  • Trace Impurities - Neodymium (Nd):
    ≤0.5 ppb
  • Trace Impurities - Nickel (Ni):
    ≤1 ppb
  • Trace Impurities - Niobium (Nb):
    max. 0.5
  • Trace Impurities - Palladium (Pd):
    ≤0.5 ppb
  • Trace Impurities - Platinum (Pt):
    ≤0.5 ppb
  • Trace Impurities - Potassium (K):
    ≤1 ppb
  • Trace Impurities - Praseodymium (Pr):
    ≤0.5 ppb
  • Trace Impurities - Rhodium (Rh):
    ≤0.5 ppb
  • Trace Impurities - Rubidium (Rb):
    ≤0.5 ppb
  • Trace Impurities - Samarium (Sm):
    ≤0.5 ppb
  • Trace Impurities - Scandium (Sc):
    ≤0.5 ppb
  • Trace Impurities - Silver (Ag):
    ≤1 ppb
  • Trace Impurities - Sodium (Na):
    ≤1 ppb
  • Trace Impurities - Strontium (Sr):
    ≤0.5 ppb
  • Trace Impurities - Tantalum (Ta):
    max. 0.5
  • Trace Impurities - Tellurium (Te):
    ≤0.5 ppb
  • Trace Impurities - Terbium (Tb):
    ≤0.5 ppb
  • Trace Impurities - Thallium (Tl):
    ≤0.5 ppb
  • Trace Impurities - Thorium (Th):
    ≤1 ppb
  • Trace Impurities - Thulium (Tm):
    ≤0.5 ppb
  • Trace Impurities - Tin (Sn):
    ≤1 ppb
  • Trace Impurities - Titanium (Ti):
    ≤1 ppb
  • Trace Impurities - Tungsten (W):
    max. 0.5
  • Trace Impurities - Uranium (U):
    ≤0.5 ppb
  • Trace Impurities - Vanadium (V):
    ≤0.5 ppb
  • Trace Impurities - Ytterbium (Yb):
    ≤0.5 ppb
  • Trace Impurities - Yttrium (Y):
    ≤0.5 ppb
  • Trace Impurities - Zinc (Zn):
    ≤1 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤0.5 ppb

Specification Test Results

Related Information

Frequently Bought Together