Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP), Finyte® for microelectronic, J.T.Baker®
N-Methyl-2-pyrrolidone (NMP), Finyte® for microelectronic, J.T.Baker®
Catalog # 6347-09
Supplier:  Avantor
CAS Number:  
undefined
N-Methyl-2-pyrrolidone (NMP), Finyte® for microelectronic, J.T.Baker®
Catalog # 6347-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    460 lb
  • Seq:
    7308

Specifications

Specification Test Results

  • Assay (C₅H₉NO):
    ≥99.8 %
  • Color (APHA):
    ≤30
  • Water (H₂O)(by Karl Fischer titrn):
    ≤300 ppm
  • Free Amines (as CH₃NH₂):
    ≤50 ppm
  • Chloride (Cl):
    ≤0.5 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Trace Impurities - Aluminum (Al):
    ≤10.0 ppb
  • Arsenic and Antimony (as As):
    ≤10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤10.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb
  • Particle Count at point of fill - 0.5 µm and greater (Rion KS42AF):
    ≤400 par/ml
  • Particle Count at point of fill - 1.0 µm and greater (Rion KS42AF):
    ≤25 par/ml

Specification Test Results

About this item

 :
 :99.13 g/mol
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193