Order Entry
Belgium
ContactUsLinkComponent
n-Hexane, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
n-Hexane, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 8473.2500
Supplier:  Avantor
CAS Number:  
undefined
n-Hexane, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 8473.2500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.0 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Water (KF):
    max. 0.02 %

Specification Test Results

About this item

 :
 :86.18 g/mol
UN:1208
ADR:3,II
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520