Order Entry
Belgium
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.9%, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
N,N-Dimethylformamide ≥99.9%, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
Catalog # 9753.1000
Supplier:  Avantor
CAS Number:  
undefined
N,N-Dimethylformamide ≥99.9%, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
Catalog # 9753.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.9 %
  • Water (by KF):
    max. 0.03 %
  • Refractive Index n20/D:
    Dependent on chemical
  • Acidity:
    max. 0.0005 meq/g
  • Residue after Evaporation:
    max. 0.0005 %
  • GC-HS Suitability:
    passes test
  • Benzene:
    indeterminable
  • Carbon tetrachloride:
    indeterminable
  • 1,2-Dichloroethane:
    indeterminable
  • 1,1-Dichloroethane:
    indeterminable
  • 1,1,1-Trichloroethane:
    indeterminable
  • Acetonitrile:
    max. 4 ppm
  • Cyclohexane:
    max. 1 ppm
  • Dichloromethane:
    max. 0.6 ppm
  • 1,2-Dimethoxyethane:
    max. 5 ppm
  • 1,4-Dioxane:
    max. 4 ppm
  • Hexane:
    max. 0.3 ppm
  • Methanol:
    max. 1 ppm
  • Methylcyclohexane:
    max. 1 ppm
  • Pyridine:
    max. 1 ppm
  • Tetrahydrofuran:
    max. 0.7 ppm
  • Toluene:
    max. 0.9 ppm
  • p-xylen:
    max. 1 ppm
  • m-Xylen:
    max. 1 ppm
  • o-xylene:
    max. 1 ppm
  • 3 Class solvents residue in total::
    max. 0.5 %
  • Acetone:
    max. 1 ppm
  • 1-Butanol:
    max. 1 ppm
  • Butyl acetate:
    max. 1 ppm
  • Ethanol:
    max. 1 ppm
  • Ethyl acetale:
    max. 1 ppm
  • Heptane:
    max. 1 ppm
  • Iso-propyl acetate:
    max. 1 ppm
  • 1-Propanol:
    max. 1 ppm
  • 2-Propanol:
    max. 1 ppm

Specification Test Results

About this item

 :
 :73.09 g/mol
UN:2265
ADR:3,III
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284