Order Entry
Belgium
ContactUsLinkComponent
Methanol, anhydrous ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Flammable
Toxic
Health
Methanol, anhydrous ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6712-25
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol, anhydrous ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6712-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    ≥ 99.8 %
  • Water (KF) :
    ≤ 0.05 %
  • Titrable Acid (meq/g) :
    ≤ 0.0005
  • Titrable Base (meq/g) :
    ≤ 0.0005
  • Residue after Evaporation :
    ≤ 0.0005 %
  • Color (APHA) :
    ≤ 10
  • Ultraviolet Absorbance (1.00-cm cell vs. water)::
    0
  • 210 nm :
    ≤ 0.6
  • 220 nm :
    ≤ 0.3
  • 230 nm :
    ≤ 0.2
  • 240 nm :
    ≤ 0.05
  • 260 nm :
    ≤ 0.01

Specification Test Results

About this item

 :
 :32.04 g/mol
Storage Temperature:Ambient
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595