Order Entry
Belgium
ContactUsLinkComponent
Manganese(II) chloride tetrahydrate 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Environment
Manganese(II) chloride tetrahydrate 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Environment
Manganese(II) chloride tetrahydrate 98.0-101.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 101.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn):
    98.0 - 101.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    3.5 - 6.0
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Potassium (K):
    ≤0.01 %
  • ACS - Sodium (Na):
    ≤0.05 %
  • ACS - Zinc (Zn):
    ≤0.005 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together