Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium sulphate heptahydrate 99-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™
 
undefined
Magnesium sulphate heptahydrate 99-100.5%, GenAR® USP for parenteral use, Macron Fine Chemicals™
 

 

  • Food GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Food Chemical:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (as MgSO₄):
    99.0 - 100.5 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - pH (1 in 20):
    5.0 - 8.8
  • USP - Loss on Ignition:
    40.0 - 52.0 %
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Selenium (Se):
    ≤ 0.003 %
  • USP - Iron (Fe)(µg/g):
    ≤ 0.5
  • Solubility (5 in 50):
    Passes Test
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    99.5 - 100.5 %
  • Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Loss on Ignition:
    40.0 - 52.0 %
  • FCC - Lead (Pb):
    = 4 mg/kg
  • FCC - Selenium (Se):
    = 0.003 %

 

Frequently Bought Together