Order Entry
Belgium
ContactUsLinkComponent
L(+)-Lysine monohydrochloride 98.5-101.5% (dried basis), GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
L(+)-Lysine monohydrochloride 98.5-101.5% (dried basis), GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
Certificates
undefined
L(+)-Lysine monohydrochloride 98.5-101.5% (dried basis), GenAR® USP, Multi-Compendial, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets J.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • Meets U.S.P Requirements:
  • GMP Manufactured Product:
  • Meets B.P. Chemical Specifications:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    20.4 - 21.4 Degree
  • USP - Loss on Drying at 105°C:
    ≤0.4 %
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Sulfate (SO₄):
    ≤0.03 %
  • USP - Iron (Fe):
    ≤0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤2.0 %
  • USP - Chloride (Cl):
    19.0 - 19.6 %
  • EP/BP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    21.0 - 22.5 Degree
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤1.0 %
  • EP/BP - Sulfate (SO₄):
    ≤300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤0.02 %
  • EP/BP - Iron (Fe):
    ≤30 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.5 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JP - Assay (C₆H₁₄N₂O₂·HCl) (dried basis):
    98.5 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Optical Rotation (+):
    19.0 - 21.5 Degree
  • JP - pH (1 in 10):
    5.0 - 6.0
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤0.028 %
  • JP - Ammonium (NH₄):
    ≤0.02 %
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤1.0 %
  • JP - Residue on Ignition:
    ≤0.10 %
  • Endotoxin Concentration, IU/mg, For Information Only:
    19.0 - 21.5 Degree
  • No Class 1,2,3 or other solvents are used or produced in the:
    5.0 - 6.0
  • manufacturing or purification of the product.:
    Passes Test
  • Elemental Impurities (USP (232), EP 5.20) - Information on elemental:
    = 0.02 %
  • impurities for this product is available on the associated Product:
    = 10 ppm
  • Regulatory Data Sheet and elemental impurity profile report.:
    = 2 ppm
  • Endotoxin Concentration, IU/mg:
    For Information Only

Specification Test Results

Related Information

Frequently Bought Together