Order Entry
Belgium
ContactUsLinkComponent
Lactic acid 88.0-92.0%, Multi-Compendial, J.T.Baker®
Corrosive
Lactic acid 88.0-92.0%, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Lactic acid 88.0-92.0%, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets E.P. Chemical Specifications:
    88.0 - 92.0%
  • GMP Manufactured Product:
    Passes Test
  • Meets U.S.P Requirements:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (CH₃CHOHCOOH):
    88.0 - 92.0 %
  • USP - Chloride (Cl):
    Passes Test
  • USP - Citric, Oxalic, Phosphoric, or Tartaric Acids:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Readily Carbonizable Substances:
    Passes Test
  • USP - Residue on Ignition:
    ≤ 0.05 %
  • USP - Sugars:
    Passes Test
  • USP - Sulfate (SO₄):
    Passes Test
  • EP - Assay (CH₃CHOHCOOH):
    88.0 - 92.0 %
  • EP - Appearance:
    Passes Test
  • EP - (S)-enantiomer:
    ≥ 95.0 %
  • EP - Calcium (Ca):
    ≤ 200 ppm
  • EP - Citric, Oxalic, and Phosphoric Acids:
    Passes Test
  • EP - Endotoxin Concentration, <5 IU/g:
    Passes Test
  • EP - Ether-Insoluble Substances:
    Passes Test
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Methanol (by GC):
    ≤ 50 ppm
  • EP - Sugars and Other Reducing Substances:
    Passes Test
  • EP - Sulfate (SO₄):
    ≤ 200 ppm
  • EP - Sulfated Ash:
    ≤ 0.1 %
  • JP - Assay (CH₃CHOHCOOH):
    85.0 - 92.0 %
  • JP - Chloride (Cl):
    ≤ 0.036 %
  • JP - Citric, Oxalic, Phosphoric, or Tartaric Acids:
    Passes Test
  • JP - Cyanide (CN):
    Passes Test
  • JP - Glycerin and Mannitol:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Identification:
    Passes Test
  • JP - Iron (Fe):
    ≤ 5 ppm
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Residue on Ignition:
    ≤ 0.10 %
  • JP - Sugars:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤ 0.010 %
  • JP - Volatile Fatty Acids:
    Passes Test
  • JP - Optical Rotation:
    -52 - -46 Degree

Specification Test Results

Related Information

Frequently Bought Together