Order Entry
Belgium
ContactUsLinkComponent
L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, J.T.Baker®
L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, J.T.Baker®
  2081-07
 :  Avantor
 :  
undefined
L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, J.T.Baker®
  2081-07
 :  Avantor
 :  

 

  • Pack type:
    HDPE pail, Flowmor™
  • Pk:
    12 kg
  • Seq:
    4187

 

 

  • EP/BP - Assay (C₆H₉N₃O₂·HCl) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Identification F:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    3.0 - 5.0
  • EP/BP - Specific Rotation [Å]²º^D (+):
    9.2 - 10.6 Degree
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤0.5 %
  • EP/BP - Sulfate (SO₄):
    ≤300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤0.02 %
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Drying at 150°C:
    7.0 - 10.0 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation (+):
    9.2 - 10.6
  • JP - pH:
    3.5 - 4.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤0.028 %
  • JP - Ammonium (NH₄):
    ≤0.02 %
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Iron (Fe):
    ≤10 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Water:
    7.2 - 10.0 %
  • JP - Residue on Ignition:
    ≤0.10 %
  • JP - Assay (anhydrous basis):
    99.0 - 101.0 %
  • Endotoxin Concentration, IU/mg ,For Information Only:
    ≤ 0.10 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    99.0 - 101.0 %
  • manufacturing or purification of the product.:
    ≤ 0.02 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    ≤ 10 ppm
  • Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
    ≤ 10 ppm
  • impurities for this product is available on the associated Product:
    Passes Test
  • Regulatory Data Sheet and elemental impurity profile report.:
    7.2 - 10.0 %

 

 

 :
 :209.63 g/mol
 :13,04741
 :259 °C
 :1.485 g/cm³ (20 °C)
 :Ambient
 :211-438-9
 :5934-29-2
 :C₆H₉N₃O₂·HCl·H₂O
 :MFCD00151027