Order Entry
Belgium
ContactUsLinkComponent
Hydrogen peroxide 30%, Finyte® for microelectronic, J.T.Baker®
Oxidizing
Irritant
Corrosive
Toxic
Health
Hydrogen peroxide 30%, Finyte® for microelectronic, J.T.Baker®
Catalog # 5846-03
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Irritant
Corrosive
Toxic
Health
Hydrogen peroxide 30%, Finyte® for microelectronic, J.T.Baker®
Catalog # 5846-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    8 pt US
  • Seq:
    8026

Specifications

Specification Test Results

  • For Microelectronic Use:
    30.0 - 32.0 %
  • Assay (H₂O₂):
    30.0 - 32.0 %
  • Color (APHA):
    ≤ 6
  • Free Acid (µeq/g):
    ≤ 0.6
  • Residue after Evaporation:
    ≤ 5 ppm
  • Chloride (Cl):
    ≤ 0.06 ppm
  • Nitrate (NO₃):
    ≤ 0.4 ppm
  • Phosphate (PO₄):
    ≤ 0.20 ppm
  • Sulfate (SO₄):
    ≤ 0.2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 10.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 10.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 10.0 ppb
  • Particle Count - 0.2 µm and greater:
    ≤ 1175 par/ml
  • Particle Count - 0.5 µm and greater:
    ≤ 25 par/ml

Specification Test Results

About this item

 :
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
UN:2014
ADR:5.1,II
 :Management of Change (MOC) category = R
 :34.01 g/mol
EINECS:231-765-0
 :GHS03
 :GHS07
 :GHS05
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333