Order Entry
Belgium
ContactUsLinkComponent
 
 
Guanidinium hydrochloride ≥99.5%, GenAR® for biotechnology, Macron Fine Chemicals™
 
undefined
 
Guanidinium hydrochloride ≥99.5%, GenAR® for biotechnology, Macron Fine Chemicals™
 

 

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (NH₂C(:NH)NH₂·HCl):
    ≥99.5 %
  • Acidity (as HCl):
    ≤0.01 %
  • Copper (Cu):
    ≤10 ppm
  • Heavy Metals (as Pb):
    ≤10 ppm
  • Iron (Fe):
    ≤10 ppm
  • Loss on Drying at 105°C:
    ≤0.5 %
  • Enzyme Activity - DNase Activity:
    Passes Test
  • Enzyme Activity - RNase Activity:
    Passes Test
  • Enzyme Activity - Protease Activity:
    Passes Test
  • Absorbance of a 6M Solution (260 nm) (au):
    ≤0.030
  • Residue after Ignition:
    ≤0.20 %

 

 

Frequently Bought Together