Order Entry
Belgium
ContactUsLinkComponent
Guanidinium hydrochloride, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidinium hydrochloride, GenAR® for biotechnology, Macron Fine Chemicals™
Catalog # 0834-26
Supplier:  Avantor
CAS Number:  
undefined
Guanidinium hydrochloride, GenAR® for biotechnology, Macron Fine Chemicals™
Catalog # 0834-26
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    200 L
  • Seq:
    5549

Specifications

About this item

 :
 :95.53 g/mol
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026