Order Entry
Belgium
ContactUsLinkComponent
Formic acid 88%, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Flammable
Irritant
Formic acid 88%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0128-01
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Flammable
Irritant
Formic acid 88%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0128-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Pk Info:
    500 ml

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥88.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤15
  • Assay (HCOOH):
    ≥88.0 %
  • Color (APHA):
    ≤15
  • Dilution Test:
    Passes Test
  • Residue after Evaporation:
    ≤0.0020 %
  • Acetic Acid (CH₃COOH):
    ≤0.4 %
  • Ammonium (NH₄):
    ≤0.005 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Sulfite (SO₃):
    Passes Test
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :46.03 g/mol
Merck Index:13,04265
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8, 3,II
 :GHS05
 :GHS02
 :GHS07
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297