Order Entry
Belgium
ContactUsLinkComponent
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Irritant
Health
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Irritant
Health
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • Assay (HOCH₂CH₂OH) (by GC):
    ≥99 %
  • Assay (HOCH₂CH₂OH)(wet analysis):
    ≥99 %
  • Color (APHA):
    ≤10
  • Specific Gravity at 20°/20°C:
    1.114 - 1.116
  • Residue after Ignition:
    ≤0.005 %
  • Acidity (as CH₃COOH):
    ≤0.01 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.2 %
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.2 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together