Order Entry
Belgium
ContactUsLinkComponent
Ethyl acetate ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Flammable
Irritant
Health
Ethyl acetate ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # 4992-26
CAS Number:  
undefined
Flammable
Irritant
Health
Ethyl acetate ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # 4992-26
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    ≥99.5%
  • ACS - Assay (CH₃COOC₂H₅) (by GC, corrected for water):
    ≥99.5 %
  • Appearance (clear, colorless liquid):
    Passes Test
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.003 %
  • ACS - Solubility in H₂O:
    Passes Test
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test
  • ACS - Titrable Acid (meq/g):
    ≤0.0009
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.2 %

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:13,03792
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
 :GHS08
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171