
Ethanol absolute ≥99.5%, BAKER ANALYZED® for UV spectrophotometry, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Storage Temperature:Ambient
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Assay:min. 99.5 %
- Residue after Evaporation:max. 0.001 %
- Water (H2O):max. 0.2 %
- at 210 nm:max. 1.00
- at 220 nm:max. 0.50
- at 230 nm:max. 0.20
- at 250 nm:max. 0.05
- at 270-400 nm:max. 0.01
Specification Test Results
Frequently Bought Together