Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate, Macron Fine Chemicals™
Disodium hydrogen phosphate dodecahydrate, Macron Fine Chemicals™
Catalog # P103-43
CAS Number:  
Disodium hydrogen phosphate dodecahydrate, Macron Fine Chemicals™
Catalog # P103-43
CAS Number:  

Specifications

  • Pk:
    35 kg
  • Seq:
    4343

Specifications

About this item

 :
 :358.14 g/mol
CAS Number:10039-32-4
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181