Order Entry
Belgium
ContactUsLinkComponent
 
Dimethyl sulphoxide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
  9754.1000
 :  Avantor
   Safer Human and Environmental Health
 :  
undefined
Dimethyl sulphoxide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
  9754.1000
 :  Avantor
   Safer Human and Environmental Health
 :  

 

  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

 

 

  • Assay (by GC):
    min. 99.9 %
  • Acidity (meq/g):
    max. 0.0005
  • Evaporation Residue:
    max. 0.0005 %
  • GC-HS Suitability:
    passes test
  • Water (KF):
    max. 0.05 %

 

 

 :
 :78.14 g/mol
 :67-68-5
 :(CH₃)₂SO
 :MFCD00002089