Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Flammable
Irritant
Diethyl ether stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 8467.2500
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Diethyl ether stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 8467.2500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.5 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Preservative (Ethanol):
    about 1%
  • Water (KF):
    max. 0.1 %

Specification Test Results

About this item

 :
 :74.12 g/mol
ADR:3,I
 :Stabilised with ethanol ∼ 1%
UN:1155
 :GHS02
 :GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646