Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, ChromAR® for HPLC, Macron Fine Chemicals™
Diethyl ether, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6727-25
CAS Number:  
undefined
Diethyl ether, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6727-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :74.12 g/mol
UN:1155
ADR:3,I
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646