Order Entry
Belgium
ContactUsLinkComponent
 
Health
Irritant
Dichloromethane ≥99.8% stabilised, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6714-25
CAS Number:  
undefined
Health
Irritant
Dichloromethane ≥99.8% stabilised, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6714-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC) :
    ≥99.8%
  • Water (KF) :
    ≤ 0.02 %
  • Titrable Acid (meq/g) :
    ≤ 0.0005
  • Residue after Evaporation :
    ≤ 0.0005 %
  • 235 nm :
    ≤ 0.5
  • 240 nm :
    ≤ 0.2
  • 250 nm :
    ≤ 0.07
  • 260 nm :
    ≤ 0.01
  • UV Cut-off :
    ≤ 229 nm
  • Stabiliser (Amylene) :
    ~50 ppm

Specification Test Results

About this item

 :
 :84.93 g/mol
Boiling Pt:39.8 °C (1013 hPa)
Storage Temperature:Ambient
UN:1593
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881