Order Entry
Belgium
ContactUsLinkComponent
Cyclohexanone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Flammable
Toxic
Health
Cyclohexanone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Catalog # 6787-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Toxic
Health
Cyclohexanone, AR®, Laboratory Reagent, Macron Fine Chemicals™
Catalog # 6787-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
 :98.14 g/mol
Boiling Pt:154.3…155.6 °C
Melting Pt:–31…–28.9 °C
Flash Pt:43…44 °C (closed cup)
UN:1145
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:108-94-1
Formula:C₆H₁₀(=O)
MDL Number:MFCD00001625