Citric acid monohydrate 99.0-102.0% (by titrimetric analysis), granular, AR® ACS, Macron Fine Chemicals™
: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
:5949-29-1
:
:C₆H₈O₇.H₂O
:MFCD00149972
:210.14 g/mol
:13,02350
:135…152 °C
:1.54 g/cm³ (20 °C)
:173.9 °C (closed cup)
:201-069-1
:GHS07
:Ambient
- Environmentally Preferable:
- Meets ACS Reagent Chemical Requirements:99.0 - 102.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- ACS - Assay (C₆H₈O₇·H₂O) (by titration):99.0 - 102.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Residue after Ignition:≤0.02 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Oxalate (C₂O₄):Passes Test
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Sulfur Compounds (as SO₄):≤0.002 %
- ACS - Iron (Fe):≤3 ppm
- ACS - Lead (Pb):≤2 ppm
- ACS - Substances Carbonizable by Hot H₂SO₄:Passes Test
Frequently Bought Together