Order Entry
Belgium
ContactUsLinkComponent
1-Butanol ≥99.4% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Flammable
Health
Irritant
1-Butanol ≥99.4% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # 3000-26
CAS Number:  
undefined
Flammable
Health
Irritant
1-Butanol ≥99.4% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Catalog # 3000-26
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.4 %
  • ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water):
    ≥99.4 %
  • ACS - Butyl Ether (C₈H₁₈O)(by GC):
    ≤0.2 %
  • ACS - Carbonyl Compounds (as Butyraldehyde):
    ≤0.01 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Evaporation:
    ≤0.005 %
  • ACS - Titrable Acid (as CH₃COOH) (meq/g):
    ≤0.0008
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.1 %

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:37 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
 :GHS02
 :GHS08
 :GHS07
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964