Order Entry
Belgium
ContactUsLinkComponent
L(+)-Ascorbic acid, fine powder USP, Macron Fine Chemicals™
L(+)-Ascorbic acid, fine powder USP, Macron Fine Chemicals™
Catalog # 8829-03
CAS Number:  
undefined
L(+)-Ascorbic acid, fine powder USP, Macron Fine Chemicals™
Catalog # 8829-03
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    99.0 - 100.5 %
  • USP - Assay (C₆H₈O₆):
    99.0 - 100.5 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Specific Rotation, [Å]²⁵^D (c=10 in water)(+):
    20.5 - 21.5 Degree

Specification Test Results

About this item

 :
 :176.13 g/mol
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328