Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 99.0-100.5% NF, ACS, J.T.Baker®
Health
Ammonium sulphate 99.0-100.5% NF, ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Health
Ammonium sulphate 99.0-100.5% NF, ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    99.0 - 100.5%
  • Meets N.F. Requirements:
    ≤0.005%
  • Meets ACS Reagent Chemical Requirements:
    ≤0.005%
  • CAUTION: For Manufacturing, processing or repackaging:
    5.0 - 6.0
  • Bulk Pharmaceutical Chemical:
    ≤0.001%
  • NF - Assay ((NH₄)₂SO₄):
    99.0 - 100.5 %
  • NF - Insoluble Matter:
    ≤0.005 %
  • NF - Residue on Ignition:
    ≤0.005 %
  • NF - pH of 5% Solution:
    5.0 - 6.0
  • NF - Nitrate (NO₃):
    ≤0.001 %
  • NF - Chloride (Cl):
    ≤ 5 ppm
  • NF - Phosphate (PO₄):
    ≤ 5 ppm
  • NF - Iron (Fe):
    ≤ 5 ppm
  • NF - Identification:
    Passes Test
  • NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g):
    ≤ 1000
  • NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g):
    ≤ 10
  • Endotoxin Concentration (EU/g):
    ≤2.5
  • Nickel (Ni):
    ≤0.7 ppm
  • Cobalt (Co):
    ≤0.09 ppm
  • Chromium (Cr):
    ≤0.03 ppm
  • Copper (Cu):
    ≤3.45 ppm
  • Manganese (Mn):
    ≤ 3.45 ppm
  • Zinc (Zn) (For Information Only):
    ≤5 ppm
  • Ni / Zn Ratio (For Information Only):
    ≤5 ppm
  • Co / Zn Ratio (For Information Only):
    ≤5 ppm
  • Cu / Zn Ratio (For Information Only):
    ≥99.0 %
  • ACS - Assay ((NH₄)₂SO₄) (A.C.S.):
    ≤0.005 %
  • ACS - Residue on Ignition:
    ≤ 0.005 %
  • ACS - pH of 5% Solution at 25°C:
    ≤0.005 %
  • ACS - Insoluble Matter:
    ≤ 0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Nitrate (NO₃):
    ≤ 0.001 %
  • ACS - Phosphate (PO₄):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • ACS - Iron (Fe):
    ≤ 5 ppm
  • Solution Test:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together