Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Health
Ammonium sulphate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Catalog # 3496-28
CAS Number:  
undefined
Health
Ammonium sulphate 99.0-100.5%, granular FCC, purified, Macron Fine Chemicals™
Catalog # 3496-28
CAS Number:  

Specifications

  • Pack type:
    Fibre drum
  • Pk:
    250 lb
  • Seq:
    7215

Specifications

Specification Test Results

  • Bulk Food Chemical:
    99.0 - 100.5 %
  • FCC - Assay ((NH₄)₂SO₄):
    99.0 - 100.5 %
  • FCC - Arsenic (As):
    ≤3 ppm
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Iron (Fe):
    ≤0.0010 %
  • FCC - Lead (Pb):
    ≤3 mg/kg
  • FCC - pH of Aqueous Solution:
    4.5 - 6.0
  • FCC - Residue after Ignition:
    ≤0.25 %
  • FCC - Selenium (Se):
    ≤0.0030 %
  • Solubility:
    Passes Test

Specification Test Results

About this item

 :
 :132.14 g/mol
Density:1.77 g/cm³
 :GHS08
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391