Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Ammonium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Ammonium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • ACS - Assay ((NH₄)₂SO₄) (A.C.S.):
    ≥99.0 %
  • ACS - Residue on Ignition:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 6.0
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤3 ppm
  • ACS - Nitrate (NO₃):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤3 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • Heavy Metals (as Pb):
    ≤3 ppm
  • Arsenic (As):
    ≤0.200 ppm

Specification Test Results

Related Information

Frequently Bought Together