Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate ≥99.0%, GenAR® for biotechnology, Macron Fine Chemicals™
Ammonium sulphate ≥99.0%, GenAR® for biotechnology, Macron Fine Chemicals™
Certificates
undefined
Ammonium sulphate ≥99.0%, GenAR® for biotechnology, Macron Fine Chemicals™
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay:
    ≥99.0 %
  • Solubility:
    Passes Test
  • ACS - Arsenic (As):
    ≤0.00002 %
  • ACS - Chloride (Cl):
    ≤0.0002 %
  • ACS - Heavy Metals (as Pb):
    ≤0.0005 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.0003 %
  • ACS - Nitrate (NO₃):
    ≤0.0002 %
  • ACS - Phosphate (PO₄):
    ≤0.0005 %
  • ACS - Residue on Ignition:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 6.0
  • Copper (Cu):
    ≤1 ppm
  • Zinc (Zn):
    ≤1.000 ppm
  • Enzyme Activity - DNase Activity:
    Passes Test
  • Enzyme Activity - RNase Activity:
    Passes Test
  • Enzyme Activity - Protease Activity:
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤2.5

Specification Test Results

Related Information

Frequently Bought Together