Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 6% (w/v) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Ammonium sulphate 6% (w/v) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # 0333-07
Supplier:  Avantor
CAS Number:  
undefined
Ammonium sulphate 6% (w/v) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Catalog # 0333-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Jerrican, poly pail
  • Pk:
    19 L
  • Seq:
    5282

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    5.9 - 6.1%
  • Appearance:
    Passes Test
  • Assay ((NH₄)₂SO₄) (by formol method):
    5.9 - 6.1 %
  • pH ("As Is" basis):
    5.0 - 6.0

Specification Test Results

About this item

 :
 :132.14 g/mol
Density:1.72 g/cm³
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391