Order Entry
Belgium
ContactUsLinkComponent
Ammonium sulphate 50% in aqueous solution, J.T.Baker®
Ammonium sulphate 50% in aqueous solution, J.T.Baker®
Catalog # 2652-38
Supplier:  Avantor
CAS Number:  
Ammonium sulphate 50% in aqueous solution, J.T.Baker®
Catalog # 2652-38
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Assay ((NH4)2SO4):
    min. 500 g/l
  • Density (g/ml) at 20°C:
    1.239 - 1.253
  • Escherichia coli (absent):
    none detected
  • pH at 20°C:
    6.8 - 7.2
  • Salmonella (absent):
    none detected
  • Total Aerobic Microbial Count, cfu/ml:
    max. 100
  • Total Yeast and Mold Count, cfu/ml:
    max. 100

Specification Test Results

About this item

 :
 :132.14 g/mol
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391