Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 14%, J.T.Baker®

Ammonia solution 14%, J.T.Baker®

370 lb
Catalog # 0889-15
Corrosive
Irritant
Environment
Supplier:   Avantor
Ammonia solution 14%, J.T.Baker®
Catalog # 0889-15
Supplier:  Avantor
CAS Number:  1336-21-6
Ammonia solution 14%, J.T.Baker®
Ammonia solution 14%, J.T.Baker®

370 lb

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 370 lb
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Appearance:
    Passes Test
  • Assay (as NH₃):
    11.2 - 16.8 %

Specifications

  • Pk:
    370 lb

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.