Order Entry
Belgium
ContactUsLinkComponent
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6711-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6711-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

About this item

 :
 :41.05 g/mol
UN:1648
ADR:3,II
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878