Order Entry
Belgium
ContactUsLinkComponent
 
Flammable
Irritant
Health
Acetone 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Catalog # 2462-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Health
Acetone 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Catalog # 2462-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, pressure bottom
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Specific Gravity at 25°/25°C:
    ≤0.789
  • NF - Nonvolatile Residue:
    ≤0.004 %
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Water (H₂O):
    ≤0.5 %
  • NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis):
    99.5 - 100.5 %
  • NF - Benzene:
    ≤2 ppm
  • ACS - Aldehydes (as HCHO):
    ≤0.002 %
  • ACS - Assay (CH₃COCH₃) (by GC, corrected for water):
    ≥99.5 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤0.05 %
  • ACS - Methanol (CH₃OH)(by GC):
    ≤0.05 %
  • ACS - Residue after Evaporation:
    ≤0.001 %
  • ACS - Solubility in H₂O:
    Passes Test
  • ACS - Substances Reducing Permanganate:
    Passes Test
  • ACS - Titrable Acid (meq/g):
    ≤0.0003
  • ACS - Titrable Base (meq/g):
    ≤0.0006
  • ACS - Water (H₂O):
    ≤0.5 %

Specification Test Results

About this item

 :
 :58.08 g/mol
UN:1090
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765