Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, J.T.Baker®

Acetone, J.T.Baker®

   Sustainable Options Available
Supplier:   Avantor
Acetone, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Acetone, J.T.Baker®

2 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • UN:1090
  • ADR:3,II
Specifications for Product Family

Specification Test Results

  • Assay (by GC):
    min. 99 %

Specifications

Specifications

  • Catalog No:
  • 3413.5000
  • 3413.9010
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    3413.5000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    3413.9010
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.