Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Irritant
Health
2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Health
2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • Assay (CH₃CHOHCH₃):
    ≥ 99.5 %
  • Carbonyl Compounds - Acetone:
    ≤ 0.002 %
  • Carbonyl Compounds - Propionaldehyde:
    ≤ 0.002 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 0.001 %
  • Solubility in H₂O:
    Passes Test
  • Titrable Acid or Base (meq/g):
    ≤ 0.0001
  • Water (by KF, coulometric):
    ≤ 0.2 %
  • Trace Impurities - Copper (Cu):
    ≤ 0.10 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 1 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Trace Impurities - Nickel (Ni):
    ≤ 0.1 ppm
  • Date Received: __ __ /__ __ /__ __:
  • Date Opened: __ __ /__ __ /__ __:
  • Meets Reagent Specifications for testing USP/NF monographs:

Specification Test Results

Related Information

Frequently Bought Together