Organic reference standards, fosinoprils, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:1356932-15-4
- Formula:C₃₀H₄₅NNaO₇P
- Description:Fosinopril impurity B, (2RS,4RS)-4-cyclohexyl-1-[[(RS)-[(1SR)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylic acid
Specifications
Frequently Bought Together