Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, fluvoxamines, Ph. Eur.
Organic reference standards, fluvoxamines, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, fluvoxamines, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    61718-80-7
  • Formula:
    C₁₃H₁₅F₃O₂
  • Description:
    Fluvoxamine impurity D, 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one

Specifications

Frequently Bought Together