Organic reference standards, esomeprazole magnesium trihydrate, Ph. Eur.
Catalog # EDQMY0001028
Supplier: EDQM
CAS Number:
Specifications
- Pk:5 mg
- Standards:Reference standard
- CAS No.:217087-09-7
- Description:Esomeprazole magnesium trihydrate, Ph. Eur. standard
- Formula:C₃₄H₃₆MgN₆O₆S₂
- Description:Esomeprazole magnesium trihydrate