Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, clobetasols, Ph. Eur.
Organic reference standards, clobetasols, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, clobetasols, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    25122-46-7
  • Formula:
    C₂₅H₃₂ClFO₅
  • Description:
    Clobetasol propionate

Specifications

Frequently Bought Together