Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, clioquinol, Ph. Eur.
Organic reference standards, clioquinol, Ph. Eur.
Catalog # EDQMY0000364
Supplier:  EDQM
CAS Number:  
Organic reference standards, clioquinol, Ph. Eur.
Catalog # EDQMY0000364
Supplier:  EDQM
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    Reference standard
  • CAS No.:
    130-26-7
  • Description:
    Clioquinol, Ph. Eur. standard
  • Formula:
    C₉H₅ClINO
  • Description:
    Clioquinol

Specifications

About this item