Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, anastrozole, Ph. Eur.
Organic reference standards, anastrozole, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, anastrozole, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    120511-88-8
  • Formula:
    C₁₅H₁₈N₂O
  • Description:
    Anastrozole impurity E, 2,2'-[5-(hydroxymethyl)benzene-1,3-diyl]bis(2-methylpropanenitrile)

Specifications

Frequently Bought Together