Order Entry
Belgium
ContactUsLinkComponent
Organic reference standards, acetylsalicylic acid, Ph. Eur.
Organic reference standards, acetylsalicylic acid, Ph. Eur.
Supplier:  EDQM
Certificates
Organic reference standards, acetylsalicylic acid, Ph. Eur.
Supplier:  EDQM

Specifications

  • CAS No.:
    50-78-2
  • Formula:
    C₉H₈O₄
  • Description:
    Acetylsalicylic acid for peak identification

Specifications

Frequently Bought Together