Native Bovine Fibronectin (from Plasma), Calbiochem®
Native fibronectin purified from pooled bovine plasma. Effective agent for promoting attachment of cells to commonly-used culture substrates.
- CAS No.:86088-83-7
- Protein/peptide type:Native
- Source:Plasma
- Species:Bovine
- Storage conditions:–20 °C
- Form:Liquid
- Protein/peptide name:Fibrinectin
- Purity:Purified from pooled bovine plasma
- Molecular weight:440,000
- Formulation:In 150 mM NaCl, 20 mM sodium phosphate, pH 7.3
Frequently Bought Together